• USA Home
  • N9648 - Anti-Neurotensin antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
N9648 Sigma

Anti-Neurotensin antibody produced in rabbit

whole antiserum, lyophilized powder

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Neurobiology, Antibodies to Neural Proteins, NE-NI,
species reactivity   rat
application(s)   immunohistochemistry: suitable
  indirect immunofluorescence: suitable
clone   polyclonal
antibody form   whole antiserum
form   lyophilized powder
shipped in   wet ice
storage temp.   −20°C
Gene Information   rat ... Nts(299757)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic neurotensin

Antigen Background

Neurotensin a 13 amino acid (ELYENKPRRPYIL) peptide, has potent neuropharmacological properties including analgesia, hypothermia, accentuation of barbiturate and ethanol sleeping time and increased release of growth hormone and prolactin.

Price and Availability


Detect Your Unseen Protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?