• USA Home
  • N9750 - α-Naphthyl nonanoate

EMAIL THIS PAGE TO A FRIEND
N9750 Sigma

α-Naphthyl nonanoate

~95%

Synonym: Nonanoic acid 1-naphthyl ester

Purchase

Properties

assay   ~95%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?