• USA Home
  • N9888 - Nitrocellulose membranes for hybridization

EMAIL THIS PAGE TO A FRIEND
N9888 Sigma

Nitrocellulose membranes for hybridization

pore size 0.45 μm, size 20 cm × 20 cm

Purchase

Properties

Related Categories Hybridization Membranes, Hybridization Supplies, Labware, Molecular Biology Supplies
size   20 cm × 20 cm
pore size   0.45 μm

Description

Application

For use in dot blots and Southern blots

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3270 4.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?