• USA Home
  • N9905 - Neuropeptide SF human

EMAIL THIS PAGE TO A FRIEND
N9905 Sigma

Neuropeptide SF human

≥92% (HPLC), solid

  • CAS Number 192387-39-6

  • Empirical Formula (Hill Notation) C65H94N18O15

  • Molecular Weight 1367.55

  •  MDL number MFCD03093437

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Neuropeptides, Opioid Peptides, Other Opioid Peptides,
InChI Key   ZOCAYXQBGJIAOK-UDXVSVLKSA-N
assay   ≥92% (HPLC)
form   solid
storage temp.   −20°C
Gene Information   human ... NPFF(8620)

Description

Amino Acid Sequence

Ser-Gln-Ala-Phe-Leu-Phe-Gln-Pro-Gln-Arg-Phe-NH2

Biochem/physiol Actions

Anti-opiate neuropeptide implicated in pain modulation.

Price and Availability


Making Headway

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?