• USA Home
  • N9908 - [Phe1-ψ(CH2-NH)-Gly2]-Nociceptin Fragment 1-13 amide

EMAIL THIS PAGE TO A FRIEND
N9908 Sigma

[Phe1-ψ(CH2-NH)-Gly2]-Nociceptin Fragment 1-13 amide

>95%

  • Empirical Formula (Hill Notation) C61H101N22O14

  • Molecular Weight 1366.59

  •  MDL number MFCD01631267

  •  PubChem Substance ID 329818698

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, N,
InChI Key   ZHKMSRDIVOXQKP-YILJZHMHSA-N
assay   >95%
storage temp.   −20°C
Gene Information   human ... PNOC(5368)

Description

Amino Acid Sequence

Phe-(CH2-NH)-Gly-Gly-Phe-Thr-Gly-Ala-Arg-Lys-Ser-Ala-Arg-Lys-NH2

Biochem/physiol Actions

Selective ORL1 opioid receptor antagonist at peripheral receptors. It functions as a partial agonist at central ORL1 receptors.

Features and Benefits

This compound is featured on the Opioid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Price and Availability


Better Together

Discover your Target

Making Headway
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?