EMAIL THIS PAGE TO A FRIEND
NSR3C Sigma

CED Phosphoramidites

Purchase

Properties

storage temp.   −20°C

Description

Packaging

Contains each of four N-blocked-5′-DMT 3′-cyanoethyl diisopropyl phosphoramidites

Price and Availability

Standard component

Description

Product #

Add to Cart

N-blocked-5′-DMT 3′-cyanoethyl diisopropyl phosphoramidite ,T 1 mL/vial SDS    
N-blocked-5′-DMT 3′-cyanoethyl diisopropyl phosphoramidite, N-Bz-da 1 mL/vial SDS    
N-blocked-5′-DMT 3′-cyanoethyl diisopropyl phosphoramidite, N-Bz-dC 1 mL/vial SDS    
N-blocked-5′-DMT 3′-cyanoethyl diisopropyl phosphoramidite, N-iBu-dG 1 mL/vial SDS    
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?