• USA Home
  • O005 - Nalbuphine hydrochloride

EMAIL THIS PAGE TO A FRIEND
O005 Sigma

Nalbuphine hydrochloride

Synonym: 17-(Cyclobutylmethyl)-4,5-epoxymorphinan-3,6,14-triol

Purchase

Properties

drug control   regulated under CDSA - not available from Sigma-Aldrich Canada

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
QD3181000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

N4396 Nalbuphine hydrochloride hydrate, ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?