EMAIL THIS PAGE TO A FRIEND
O0639 Sigma
Nunc® OmniTray
Maxisorp with lid
Purchase
Properties
Related Categories Genomics, PCR and Arrays, Labware, Manufacturer Browser, Molecular Biology, Nunc,
material   polystyrene
sterility   non-sterile
  sterile
feature   lid
mfr. no.   Nunc 465219-12
packaging   case of 60
L × W   128 mm × 86 mm
capacity   90 mL
working volume   35 mL
matrix active group   non-treated surface treatment
suitability   suitable for (standard petri dish techniques and bacterial growth, screening libraries and DNA hybridization methods)
Description
General description
Nunc® OmniTray
• Ideal for screening libraries
• Useful for DNA hybridization methods
• Also suitable for standard petri dish techniques and bacterial growth
• Same external foot print dimensions as a 96 MicroWell plate
• Stackable and compatible with automatomated equipment
• Optical clarity makes colonies easy to see
• Walls are high and sturdy
• Can be frozen at -20°C
The Nunc OmniTray acts as:
• Holder for membrane when dot blotting
• Container for denaturation, hybridization and washing steps
MaxiSorp surface is ideal for arraying molecules with mixed hydrophilic/hydrophobic domains such as proteins.
Legal Information
MaxiSorp is a trademark of Thermo Fisher Scientific or its subsidiaries
MicroWell is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?