• USA Home
  • O1009 - Anti-Oncostatin M antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
O1009 Sigma

Anti-Oncostatin M antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym: Anti-OSM

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Cytokines and Growth Factors, Antibodies to Oncostatin M (OSM),
species reactivity   mouse
application(s)   indirect ELISA: suitable
  neutralization: suitable
  western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   mouse ... Osm(18413)
biological source   goat
conjugate   unconjugated

Description

Immunogen

recombinant mouse OSM expressed in Escherichia coli.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Specificity

The antibody neutralizes the biological activity of recombinant mouse OSM. It shows less than 2% cross-reactivity with recombinant human OSM.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, type N95 (US), type P1 (EN143) respirator filter
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

comparable product

Product #

Description

Add to Cart

O0884 Monoclonal Anti-Oncostatin M antibody produced in mouse, clone 17001, purified immunoglobulin, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?