• USA Home
  • O1501 - Orotidine-5′-monophosphate pyrophosphorylase from brewer's bottom yeast

EMAIL THIS PAGE TO A FRIEND
O1501 Sigma

Orotidine-5′-monophosphate pyrophosphorylase from brewer's bottom yeast

lyophilized powder, ~25 units/mg protein (modified Warburg-Christian)

Synonym: Orotidine-5′-phosphate: pyrophosphate phosphoribosyltransferase

Purchase

Properties

Related Categories 2.4.x.x Glycosyltransferases, 2.x.x.x Transferases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
form   lyophilized powder
foreign activity   OMP decarboxylase <1%
  inorganic pyrophosphatase ~5%
storage temp.   −20°C

Description

Application

This is the preferred enzyme for assaying orotidine 5′-monophosphate and for the production of OMP analogs from the corresponding orotic acid.

Unit Definition

One unit will convert 1.0 μmole of orotidine 5′-monophosphate to orotic acid in one hr at pH 8.0 at 30°C.

Physical form

Lyophilized powder containing approx. 50% buffer salts

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?