• USA Home
  • O1506 - Okadaic acid from Halichondria (black sponge)

EMAIL THIS PAGE TO A FRIEND
O1506 Sigma

Okadaic acid from Halichondria (black sponge)

≥80% (HPLC), powder

Purchase

Properties

assay   ≥80% (HPLC)
form   powder
color   white
solubility   DMSO: ≥1 mg/mL
  ethanol: ≥1 mg/mL
  methanol: ≥1 mg/mL
  H2O: insoluble
storage temp.   −20°C

Description

Physical form

Photosensitive powder

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3462 6.1 / PGI
WGK Germany 
3
RTECS 
AA8227800
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

O9381 Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?