• USA Home
  • O3001 - L-Ornithine Decarboxylase from Escherichia coli

EMAIL THIS PAGE TO A FRIEND
O3001 Sigma

L-Ornithine Decarboxylase from Escherichia coli

partially purified powder, ≥0.03 units/mg protein (biuret)

Purchase

Properties

form   partially purified powder
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?