• USA Home
  • O3006 - (S)-Oxynitrilase from Manihot esculenta (manioc)

EMAIL THIS PAGE TO A FRIEND
O3006 Sigma

(S)-Oxynitrilase from Manihot esculenta (manioc)

Purchase

Properties

InChI Key   KHLCTMQBMINUNT-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?