• USA Home
  • O3257 - Vacuum filtration manifold Millipore® model 1225 replacement parts

EMAIL THIS PAGE TO A FRIEND
O3257 Sigma

Vacuum filtration manifold Millipore® model 1225 replacement parts

Filter sealing O-rings, silicone

Purchase

Description

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1262 - class 3 - PG 2 - Octanes
WGK Germany 
2
RTECS 
RG8400000
Flash Point(F) 
55.4 °F
Flash Point(C) 
13 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?