• USA Home
  • O4382 - Osteocalcin Fragment 45-49 human

EMAIL THIS PAGE TO A FRIEND
O4382 Sigma

Osteocalcin Fragment 45-49 human

≥97% (HPLC)

Synonym: Phe-Tyr-Gly-Pro-Val

  • CAS Number 85679-70-5

  • Empirical Formula (Hill Notation) C30H39N5O7

  • Molecular Weight 581.66

  •  MDL number MFCD00076322

  •  PubChem Substance ID 24897995

Purchase

Properties

Related Categories Calcium Associated Protein Fragments, Cell Biology, Cell Signaling and Neuroscience, Ion Channel Modulating Peptides, Peptides and Proteins,
InChI Key   MJAFJCRCGVXXTP-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   human ... BGLAP(632)

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?