• USA Home
  • O4505 - Orange II sodium salt

EMAIL THIS PAGE TO A FRIEND
O4505 Sigma

Orange II sodium salt

certified by the Biological Stain Commission

Synonym: 4-(2-Hydroxy-1-naphthylazo)benzenesulfonic acid sodium salt, Acid Orange 7, Acid Orange A, Orange II, Tropaeolin 000 No. 2

  • CAS Number 633-96-5

  • Empirical Formula (Hill Notation) C16H11N2NaO4S

  • Molecular Weight 350.32

  •  Beilstein Registry Number 3898201

  •  Colour Index Number 15510

  •  EC Number 211-199-0

  •  MDL number MFCD00011657

  •  PubChem Substance ID 329818910

Purchase

Properties

Related Categories Cell Biology, Certified Stains & Dyes, Hematology and Histology, Stains and Dyes
grade   certified by the Biological Stain Commission
InChI Key   CQPFMGBJSMSXLP-ZAGWXBKKSA-M

Description

Suitability

Certified as a substitute for methyl orange in the Wilson-Ezrin method for pituitary acidophils; as a counterstain for Heidenhain′s or Weigert′s hematoxylin on paraffin sections; as a substitute for Orange G in Flemming′s triple stain.

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
DB7084000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?