• USA Home
  • O4508 - Orotate reductase from Lysobacter enzymogenes

EMAIL THIS PAGE TO A FRIEND
O4508 Sigma

Orotate reductase from Lysobacter enzymogenes

lyophilized powder, ~7.0 units/mg protein

Synonym: L-4,5-Dihydroorotate: oxygen oxidoreductase

Purchase

Properties

Related Categories 1.3.x.x Acting on CH-CH, 1.x.x.x Oxidoreductases, Biochemicals and Reagents, Enzyme Class Index, Enzymes, Inhibitors, and Substrates More...
form   lyophilized powder
composition   Protein, ~15% Lowry
storage temp.   −20°C

Description

Unit Definition

One unit will convert 1.0 μmole of orotic acid to dihydroorotate per min at pH 6.5 at 25°C.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?