• USA Home
  • O5382 - β-N-Oxalylamino-L-alanine

EMAIL THIS PAGE TO A FRIEND
O5382 Sigma

β-N-Oxalylamino-L-alanine

Synonym: L-ODAP, BOAA

Purchase

Properties

storage temp.   −20°C

Description

Biochem/physiol Actions

May be involved in the etiology of the motor neuron disease, lathyrism

Other Notes

Neuroexcitotoxic amino acid from Lathyrus sativus

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
RO4555000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?