EMAIL THIS PAGE TO A FRIEND
O5633 Sigma

d(pT)5 sodium salt

lyophilized powder

Purchase

Properties

form   lyophilized powder
storage temp.   −20°C

Description

Preparation Note

Prepared by enzymatic degradation of poly(dT) and fractionation by ion-exchange chromatography.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?