• USA Home
  • O6008 - Ovarian tumor antigen from human fluids

EMAIL THIS PAGE TO A FRIEND
O6008 Sigma

Ovarian tumor antigen from human fluids

≥50,000 units/mL, buffered aqueous solution

Synonym: Ca-125

Purchase

Properties

Related Categories Cancer Markers and Antibodies, Cancer Research, Cell Biology, Proteins and Peptides, Steroid Sensitive Cancer Markers,
form   buffered aqueous solution
concentration   ≥50,000 units/mL
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... BRCA1(672)

Description

Quality

Highly purified.

Unit Definition

One unit is an arbitrary unit related to a reference antigen preparation using CIS-RIA method.

Physical form

Solution in 0.15 M phosphate buffered saline, pH 7.4, containing 0.1% sodium azide

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?