EMAIL THIS PAGE TO A FRIEND
O6127 Sigma

Octyl–Agarose

saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Hydrophobic Interaction, Molecular Biology, Protein Chromatography, Proteomics More...
form   saline suspension
matrix activation   epoxy
matrix attachment   amino
matrix spacer   12 atoms
capacity   ≥10 mg/mL binding capacity (human serum albumin)
  ≥3 mg/mL binding capacity (β-lactoglobulin)
storage temp.   2-8°C

Description

Packaging

50 mL in glass bottle

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Application

Octyl agarose is used in protein chromatography, affinity chromatography and hydrophobic interactions. Octyl agarose has been used to immobilize lipases from Penicillium camembertii (Lipase G) and Aspergillus niger.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?