EMAIL THIS PAGE TO A FRIEND
O6137 Sigma

Orexin B human

≥97% (HPLC)

Synonym: Hypocretin 2

  • Empirical Formula (Hill Notation) C123H212N44O35S

  • Molecular Weight 2899.34

  •  MDL number MFCD01635373

Purchase

Description

Amino Acid Sequence

Arg-Ser-Gly-Pro-Pro-Gly-Leu-Gln-Gly-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Ala-Ser-Gly-Asn-His-Ala-Ala-Gly-Ile-Leu-Thr-Met-NH2

Biochem/physiol Actions

Orexins (hypocretins) comprise a family of hypothalamic peptides that are involved in the initiation of sleep and in feeding behavior. Orexin A stimulates food intake through activation of the OX1 receptor. Orexin B activates OX2 receptors. Both OX1 and OX2 receptors are involved in maintaining arousal. OX2 receptors appear to be inhibitory autoreceptors on orexin-containing neurons.

Price and Availability


Amplified Detection

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?