EMAIL THIS PAGE TO A FRIEND
O6376 Sigma

Octyl–Agarose

saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Hydrophobic Interaction, Molecular Biology, Protein Chromatography, Proteomics More...
form   saline suspension
matrix   4% beaded agarose
matrix activation   cyanogen bromide
matrix attachment   amino
matrix spacer   1 atom
capacity   ≥13 mg/mL binding capacity (bovine serum albumin)
storage temp.   2-8°C

Description

Packaging

50 mL in glass bottle

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Application

Octyl-agarose is used in protein chromatography, affinity chromatography and hydrophobic interactions. Octyl-agarose has been used to immobilize lipases from Penicillium camembertii (Lipase G) and Aspergillus niger.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?