• USA Home
  • O6633 - Components Of N-Linked Oligosaccharide Profiling Kits

EMAIL THIS PAGE TO A FRIEND
O6633 Sigma

Components Of N-Linked Oligosaccharide Profiling Kits

Purchase

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?