• USA Home
  • O6637 - BioParticles® Opsonizing Reagents

EMAIL THIS PAGE TO A FRIEND
O6637 Sigma

BioParticles® Opsonizing Reagents

Purchase

Properties

Related Categories Affinity Chromatography, Molecular Biology, Protein Chromatography, Proteomics, Reagents More...
storage temp.   −20°C

Description

Compatibility

For S. cerevisiae (Zymosan)

Application

Opsonizing reagents are used in affinity chromatography and protein chromatography. Opsonizing reagents have been used in studies to develop an easy and inexpensive method of investigating phagocytosis in retinal pigment epithelium cells. They have also been used to study roles in innate defense against infection and to investigate immunomodulating activity of medicinal plants.

Legal Information

BioParticles is a registered trademark of Life Technologies

Price and Availability


Amplified Detection

Amicon Pro Purification System
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?