• USA Home
  • O7637 - Oncyte® multiwells for cytochemistry arrays
EMAIL THIS PAGE TO A FRIEND
O7637 Sigma
Oncyte® multiwells for cytochemistry arrays
5 mm well diameter, 12 wells/slide
Purchase
Description
Compatibility
Multiwell nitrocellulose dots, formulated for fluorescent imaging, affixed to white-glass microscope slides for cytochemistry high throughput screening. Ultra-thin microporous coating insures uniform binding of cells and macromolecules. Becomes transparent for microscopy and imaging using a variety of mounting media. Removable silicon multiwells isolate specimens and reagents to prevent cross contamination. Press-to-seal silicone chamber surface seals easily to prevent evaporation. Gaskets are removed cleanly and easily, by simply peeling them off.
Legal Information
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?