EMAIL THIS PAGE TO A FRIEND
O8005 Sigma

Oxazolam

Purchase

Properties

drug control   Home Office Schedule 4.1; stupéfiant; kontrollierte Droge in Deutschlandregulated under CDSA - not available from Sigma-Aldrich Canada
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?