EMAIL THIS PAGE TO A FRIEND
O8887 Sigma

Oxonol VI

Synonym: 1,5-Bis(5-oxo-3-propylisoxazol-4-yl)pentamethine oxonol

  • CAS Number 64724-75-0

  • Empirical Formula (Hill Notation) C17H20N2O4

  • Molecular Weight 316.35

  •  MDL number MFCD00210076

Purchase

Properties

InChI Key   CPHARWHMAGUXDW-PQWGHOHWSA-N

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?