EMAIL THIS PAGE TO A FRIEND
O9012 Sigma

Oxonol V

Synonym: 1,5-Penta-1,3-dien-1-yl-5-ylidene-bis(5-oxo-3-phenyl-4-isoxazole), 4-[5-(5-Hydroxy-3-phenyl-4-isoxazolyl)-2,4-pentadienylidene]-3-phenyl-5(4H)-isoxazolone

  • CAS Number 61389-30-8

  • Empirical Formula (Hill Notation) C23H16N2O4

  • Molecular Weight 384.38

  •  Beilstein Registry Number 575970

  •  MDL number MFCD00075232

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   NEDZCQDBZLHEGJ-UFRBDIPQSA-N

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?