• USA Home
  • O9128 - d(pC)12 ammonium salt

EMAIL THIS PAGE TO A FRIEND
O9128 Sigma

d(pC)12 ammonium salt

Synonym: Oligo d(pC)n

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?