• USA Home
  • O9257 - Osteonectin from bovine bone

EMAIL THIS PAGE TO A FRIEND
O9257 Sigma

Osteonectin from bovine bone

buffered aqueous solution

Purchase

Properties

form   buffered aqueous solution
storage temp.   −20°C

Description

Packaging

Package size based on protein content

Physical form

Solution in 20 mM Tris, pH 7.4, 150 mM NaCl, 2 mM CaCl2

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?