• USA Home
  • O9756 - Anti-Orexin A antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
O9756 Sigma

Anti-Orexin A antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Obesity Research, Antibodies to Hormones,
species reactivity   rat, human, mouse
application(s)   indirect ELISA: 1:20,000
  radioimmunoassay: 1:300,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... HCRT(3060)
mouse ... Hcrt(15171)
rat ... Hcrt(25723)
biological source   rabbit
InChI Key   ALSHYECRTIFLAI-UHFFFAOYSA-N
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to the N-terminus of human orexin A (amino acids 1-17 with disulfide bridge between cysteines 6-12 and 7-14 and C-terminally added Gly-Lys), conjugated to KLH. This sequence is identical in mouse and rat orexin A and is different from the orexin B sequence.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?