• USA Home
  • P0159 - R-Phycoerythrin from Corallina officinalis

EMAIL THIS PAGE TO A FRIEND
P0159 Sigma

R-Phycoerythrin from Corallina officinalis

ammonium sulfate suspension

Synonym: PE

Purchase

Properties

Related Categories Auxiliary Proteins, Biochemicals and Reagents, Phycobiliproteins, Proteins and Derivatives
form   ammonium sulfate suspension
storage temp.   2-8°C

Description

Other Notes

Sold as mg protein based on E1%/565 = 81.7.

Packaging

Package size based on protein content

Physical form

Suspension in 50% ammonium sulfate, 50 mM sodium phosphate buffer, pH 7.0

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?