• USA Home
  • P0180 - Phosphoric acid solution

EMAIL THIS PAGE TO A FRIEND
P0180 Sigma

Phosphoric acid solution

1.0 M

Purchase

Properties

InChI Key   NBIIXXVUZAFLBC-UHFFFAOYSA-N
concentration   1.0 M

Description

Application

1M phosphoric acid is suitable as a concentrated anode solution for isoelectric focusing (IEF). A 0.1 M phosphoric acid solution is commonly used as an anode solution but some procedures use other concentrations. 1 M Phosphoric acid solution may be diluted to the desired concentration with deionized water.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1805 8 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?