• USA Home
  • P0190 - cis-10-Pentadecenoic acid

EMAIL THIS PAGE TO A FRIEND
P0190 Sigma

cis-10-Pentadecenoic acid

~98% (capillary GC)

  • CAS Number 84743-29-3

  • Empirical Formula (Hill Notation) C15H28O2

  • Molecular Weight 240.38

  •  MDL number MFCD00133179

Purchase

Properties

InChI Key   APXSAEQXOXTDAM-WAYWQWQTSA-N
assay   ~98% (capillary GC)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?