EMAIL THIS PAGE TO A FRIEND
P0230 Sigma

Poly(A)+ RNA

from mouse embryo (11-day)

Synonym: mRNA, messenger RNA

Purchase

Properties

biological source   from mouse embryo (11-day)
form   solution (in 10 mM HEPES, pH 7.5, containing 1 mM EDTA)
concentration   ~0.5 mg/mL
shipped in   dry ice

Description

General description

Sigma offers a broad range of high-quality mRNA isolated from various tissue and cell sources. Each mRNA is purified using a combination of either oligo(dT)-cellulose chromatography or proprietary isolation methods. Following purification the integrity of the mRNA is assessed using denatured agarose gel or formaldehyde gel electrophoresis followed by Northern blot analysis.
Non-human mRNA is prepared from Sprague-Dawley rats and from BALB/c mice.

Physical form

Each poly(A)+ mRNA is provided as a ready-to-use solution of 0.5 mg/ml.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?