• USA Home
  • P0242 - Hanna precision pH meter
EMAIL THIS PAGE TO A FRIEND
P0242 Sigma
Hanna precision pH meter
Model pH 209
Purchase
Properties
Related Categories Benchtop, Hanna, Hanna pH Meters, Hanna pH Meters and Accessories, Labware,
feature   calibration manual one- or two-point; offset and slope calibration settings are displayed − an excellent teaching aid
  pH calibration manual 2 points through offset & slope knobs
  temp. compensation manual from 0-100 °C
  temperature compensation 0 ‑ 100 °C (manual)
mfr. no.   Model pH 209
measuring range   −1999-1999 mV voltage, resolution: 1 mV
  0-14 pH, resolution: 0.01
  0.00-14.00 pH, accuracy: 0.01, resolution: 0.01 pH unit
  ±1999 mV mV2, accuracy: 0.1 mV, resolution: 1 mV
L × W × H   240 mm × 182 mm × 74 mm
Description
General description
Includes a 12 VDC adaptor, for Europe this will be fitted with a 2-pin Europlug.
Product does not include the electrode arm pictured.
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Rugged ABS casing with sturdy knobs and a reference chart for pH and temperature on the front panel. Measurements can be performed for pH with manual temperature compensation as well as for REDOX (Reduction Oxidation Potential).
HI 1332B (included) combination double-junction refillable pH electrode, BNC with 1 m cable included.
The keypad has membrane keys for changing the mode and knobs for slope and offset calibration. The pH offset and slope are manually adjustable to display the calibration concept and the electrode characteristics. The LCD displays the set temperature while manually compensating for its effects.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?