• USA Home
  • P0268 - Pyridine-2-aldoxime methyl methanesulfonate

EMAIL THIS PAGE TO A FRIEND
P0268 Sigma

Pyridine-2-aldoxime methyl methanesulfonate

Synonym: 2-PAM methanesulfonate salt, 2-PAM methanesulfonate, P2S, Pralidoxime mesylate salt, Pralidoxime mesylate, Pralidoxime methanesulfonate

  • CAS Number 154-97-2

  • Empirical Formula (Hill Notation) C7H9N2O · CH3O3S

  • Molecular Weight 232.26

  •  EC Number 205-839-8

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
UU4570000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?