• USA Home
  • P0279 - Parathyroid Hormone fragment 1-44 human

EMAIL THIS PAGE TO A FRIEND
P0279 Sigma

Parathyroid Hormone fragment 1-44 human

≥97% (HPLC)

  • CAS Number 85568-24-7

  • Empirical Formula (Hill Notation) C225H366N68O61S2

  • Molecular Weight 5063.86

  •  MDL number MFCD00081827

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Parathyroid Hormone Fragments, Peptides and Proteins, Peptides for Cell Biology More...
assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   human ... PTH(5741)

Description

Amino Acid Sequence

Ser-Val-Ser-Glu-Ile-Gln-Leu-Met-His-Asn-Leu-Gly-Lys-His-Leu-Asn-Ser-Met-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe-Val-Ala-Leu-Gly-Ala-Pro-Leu-Ala-Pro-Arg

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?