• USA Home
  • P0310 - Pipette tips, disposable, polypropylene, capacity 1000 μL
EMAIL THIS PAGE TO A FRIEND
P0310 Sigma
Pipette tips, disposable, polypropylene, capacity 1000 μL
With aerosol barrier, sterile
Purchase
Properties
Related Categories Labware, Liquid Handling Equipment, Miscellaneous Pipette Tips, Pipette Tips, Pipettes, Pipettors and Tips More...
sterility   sterile
packaging   tray of 96 tips
Description
General description
Inert, bonded fiber barrier coalesces airborne contaminants so they do not enter pipette barrel.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?