• USA Home
  • P0325 - Fluoropore PTFE filter membranes
EMAIL THIS PAGE TO A FRIEND
P0325 Sigma
Fluoropore PTFE filter membranes
pore size 0.2 μm, diam. 25 mm
Purchase
Properties
Related Categories Filtration Supplies, Fluoropore PTFE filter membranes, Labware, Manufacturer Browser, Membrane Filters,
mfr. no.   Millipore FGLP02500
  Millipore, FGLP02500
diam.   25 mm
pore size   0.2 μm pore size
Description
General description
Biologically inert, with broad chemical compatibility. HDPE backing to improve handling, compatible with acids, bases and solvents (except aromatic hydrocarbons above 80°C).
Legal Information
Fluoropore is a trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?