• USA Home
  • P0399 - 1,2-Dipalmitoyl-sn-glycero-3-phospho-N,N-dimethylethanolamine

EMAIL THIS PAGE TO A FRIEND
P0399 Sigma

1,2-Dipalmitoyl-sn-glycero-3-phospho-N,N-dimethylethanolamine

~99%

Synonym: 3-sn-Phosphatidyl-N,N-dimethylethanolamine, 1,2-dipalmitoyl, L-α-Phosphatidylethanolamine, dipalmitoyl, N,N-dimethyl, L-β,γ-Dipalmitoyl-N,N-dimethyl-α-cephalin

  • CAS Number 3922-61-0

  • Empirical Formula (Hill Notation) C39H78NO8P

  • Molecular Weight 720.01

  •  Beilstein Registry Number 5194073

  •  EC Number 223-494-1

  •  MDL number MFCD00066514

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Glycerophospholipid Metabolism, Glycerophospholipids, Glycerophosphonoethanolamines,
InChI Key   SKWDCOTXHWCSGS-DIPNUNPCSA-N
assay   ~99%
storage temp.   −20°C

Description

Linkage

Similar to P1634, but produced by Sigma.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?