EMAIL THIS PAGE TO A FRIEND
P0441 Sigma
Pipette workstation
Holder and bin combination
Purchase
Description
General description
A 6-position holder that fits a variety of pipettes; bin can hold either loose pipet tips or two 96-tip trays.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?