• USA Home
  • P0529 - Parathyroid Hormone Fragment 28-48 human

EMAIL THIS PAGE TO A FRIEND
P0529 Sigma

Parathyroid Hormone Fragment 28-48 human

≥95% (HPLC)

  • CAS Number 83286-22-0

  • Empirical Formula (Hill Notation) C95H150N28O29

  • Molecular Weight 2148.38

  •  MDL number MFCD00133741

  •  PubChem Substance ID 329820042

Purchase

Properties

InChI Key   INPISQDTARSRTN-DDIUPCTASA-N
assay   ≥95% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

Leu-Gln-Asp-Val-His-Asn-Phe-Val-Ala-Leu-Gly-Ala-Pro-Leu-Ala-Pro-Arg-Asp-Ala-Gly-Ser

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?