• USA Home
  • P0539 - B-Phycoerythrin pyridyldisulfide derivative from Porphyridium cruentum

EMAIL THIS PAGE TO A FRIEND
P0539 Sigma

B-Phycoerythrin pyridyldisulfide derivative from Porphyridium cruentum

lyophilized powder

Purchase

Properties

form   lyophilized powder
composition   Protein, ~20% Lowry
storage temp.   2-8°C

Description

Packaging

Package size based on protein content

Physical form

Lyophilized powder containing sucrose and sodium azide as preservatives

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?