• USA Home
  • P0541 - L-α-Phosphatidyl-D-myo-inositol 3,4,5-triphosphate, dipalmitoyl

EMAIL THIS PAGE TO A FRIEND
P0541 Sigma

L-α-Phosphatidyl-D-myo-inositol 3,4,5-triphosphate, dipalmitoyl

  • Empirical Formula (Hill Notation) C41H82O22P4

  • Molecular Weight 1050.97

  •  MDL number MFCD02259147

Purchase

Properties

solubility   DMSO: soluble
  chloroform: soluble
  ethanol: soluble
storage temp.   −20°C

Description

Biochem/physiol Actions

Signaling lipid; activates PDK1 (3-phosphoinositide-dependent protein kinase), which is necessary, but not sufficient, for the activation of PKB/Akt. PDK1 also phosphorylates and activates p70S6k.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?