• USA Home
  • P0561 - Sigma Pseudo powder explosive scent

EMAIL THIS PAGE TO A FRIEND
P0561 Sigma

Sigma Pseudo powder explosive scent

Purchase

Properties

form   neat
format   neat

Description

Application

A canine training aid for the detection of powder explosives, e.g., black, flash, smokeless, and ammonium powders.

Packaging

Each application is packaged in a flame-sealed ampule.

Legal Information

Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?