• USA Home
  • P0566 - Pipette Pump holder
EMAIL THIS PAGE TO A FRIEND
P0566 Sigma
Pipette Pump holder
overall H 15 1/2 in.
Purchase
Properties
Related Categories Labware, Liquid Handling Equipment, Pipette Controllers and Pumps More...
overall H   15 1/2 in.
Description
General description
Acrylic holder is designed to conveniently hold a pipette pump pistol grip with pipette. The pipette slips through an open slot for easy access.
Legal Information
Pipette Pump is a trademark of SP Scienceware, a division of SP
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?