EMAIL THIS PAGE TO A FRIEND
P0625 Sigma

Palmitic acid

Grade II, ~95%

Synonym: 1-Pentadecanecarboxylic acid, C16:0, Cetylic acid, Hexadecanoic acid, NSC 5030, PamOH

  • CAS Number 57-10-3

  • Linear Formula CH3(CH2)14COOH

  • Molecular Weight 256.42

  •  Beilstein Registry Number 607489

  •  EC Number 200-312-9

  •  MDL number MFCD00002747

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Acids, Acids & Bases, Biochemicals and Reagents, Chemical Synthesis, Fatty Acids and conjugates,
vapor pressure   10 mmHg ( 210 °C)
InChI Key   IPCSVZSSVZVIGE-UHFFFAOYSA-N
type   Grade II
assay   ~95%
form   powder
bp   271.5 °C/100 mmHg(lit.)
mp   61-62.5 °C(lit.)
density   0.852 g/mL at 25 °C(lit.)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
RT4550000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

27734 Palmitic acid, ≥98% palmitic acid basis (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?