• USA Home
  • P0640 - Phospholipase D from Arachis hypogaea (peanut)

EMAIL THIS PAGE TO A FRIEND
P0640 Sigma

Phospholipase D from Arachis hypogaea (peanut)

Type II, lyophilized powder, 300-700 units/mg protein

Synonym: Lecithinase D, Phosphatidylcholine phosphatidohydrolase

Purchase

Description

Analysis Note

Protein by biuret

Preparation Note

Chromatographically purified.

Unit Definition

One unit will liberate 1.0 μmol of choline from L-α-phosphatidylcholine (egg yolk) per hr at pH 5.6 at 30 °C.

Physical form

Contains buffer salts.

Biochem/physiol Actions

Hydrolyzes the phosphate bonds of phospholipids and sphingomyelin to give the corresponding phosphatidic acid.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

P0515 Phospholipase D from Arachis hypogaea (peanut), Type II, lyophilized powder, ≥60 units/mg protein

inhibitor

Product #

Description

Add to Cart

D6518 Diethylenetriaminepentaacetic acid, ≥99% (titration)
P9375 1,10-Phenanthroline monohydrate, reagent grade
E4378 Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
ED2SS Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?